4,5-Dimethoxy-2-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A285737-5G A285737-100G
77099-246EA 6 USD
77099-246 77685-814
4,5-Dimethoxy-2-nitrobenzoic acid 98%
4,5-Dimethoxy-2-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₆
MDL Number: MFCD00014697
CAS Number: 4998-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR