Solifenacin Succinate 98%

Supplier: Ambeed
Warning

Synonyms: (S)-(R)-Quinuclidin-3-yl 1-phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate succinate

A284622-250MG A284622-50MG
77099-230EA 5.31 USD
77099-230 77099-232 77099-234
Solifenacin Succinate 98%
Solifenacin Succinate

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₂·C₄H₆O₄
MW: 480.56 g/mol
MDL Number: MFCD09952287
CAS Number: 242478-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR