N-(2-Cyanophenyl)picolinamide 95%

Supplier: Ambeed
Warning

A284068-10G A284068-1G A284068-250MG A284068-25G A284068-5G
77099-218EA 293.87 USD
77099-218 77099-220 77099-222 77099-224 77099-226
N-(2-Cyanophenyl)picolinamide 95%
N-(2-Cyanophenyl)picolinamide

Precaución: for Research Use Only

Formula: C₁₃H₉N₃O
MW: 223.23 g/mol
MDL Number: MFCD02128063
CAS Number: 304650-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR