(S)-b-Amino-4-bromo-benzeneethanol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-Amino-2-(4-bromophenyl)ethanol

A283903-100MG A283903-5G A283903-1G
77099-210EA 64.72 USD
77099-210 77571-762 77685-805
(S)-b-Amino-4-bromo-benzeneethanol 97%
(S)-b-Amino-4-bromo-benzeneethanol

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MW: 216.075 g/mol
MDL Number: MFCD09256372
CAS Number: 354153-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR