3-Bromo-2-methoxy-5-nitropyridine 98%

Supplier: Ambeed
Warning

A283774-100G A283774-10G A283774-1G A283774-25G A283774-5G
77099-200EA 180 USD
77099-200 77099-202 77099-204 77099-206 77099-208
3-Bromo-2-methoxy-5-nitropyridine 98%
3-Bromo-2-methoxy-5-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O₃
MW: 233.02
MDL Number: MFCD06659517
CAS Number: 15862-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR