3-Bromo-4-chloro-5-nitropyridine 95%

Supplier: Ambeed
Warning

A282918-10G A282918-25G A282918-100G
77099-194EA 21 USD
77099-194 77099-196 77685-803
3-Bromo-4-chloro-5-nitropyridine 95%
3-Bromo-4-chloro-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClN₂O₂
MW: 237.44
MDL Number: MFCD07368836
CAS Number: 31872-63-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR