6-Chloro-3,3-dimethyl-2,3-dihydro-1H-pyrrolo[3,2-c]pyridine 95%

Supplier: Ambeed
Warning

A282330-100MG A282330-1G A282330-250MG
77099-178EA 198.39 USD
77099-178 77099-180 77099-182
6-Chloro-3,3-dimethyl-2,3-dihydro-1H-pyrrolo[3,2-c]pyridine 95%
6-Chloro-3,3-dimethyl-2,3-dihydro-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₉H₁₁ClN₂
MW: 182.65 g/mol
MDL Number: MFCD28399436
CAS Number: 1403899-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR