3-Fluorobenzonitrile 99%

Supplier: Ambeed
Danger

A280508-100G A280508-25G A280508-5G
77099-160EA 53 USD
77099-160 77099-162 77099-164
3-Fluorobenzonitrile 99%
3-Fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄FN
MDL Number: MFCD00001797
CAS Number: 403-54-3
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR