2-Amino-4-nitro-5-methoxybenzoic Acid 95%

Supplier: Ambeed
Warning

A279506-100MG A279506-1G A279506-250MG A279506-5G
77099-144EA 46 USD
77099-144 77099-146 77099-148 77901-434
2-Amino-4-nitro-5-methoxybenzoic Acid 95%
2-Amino-4-nitro-5-methoxybenzoic Acid

Precaución: for Research Use Only

Formula: C₈H₈N₂O₅
MDL Number: MFCD10568169
CAS Number: 196194-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR