2-Fluoro-6-pyridone 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-6-hydroxypyridine, 6-Fluoropyridin-2-ol

A278437-100G A278437-10G A278437-1G
77099-128EA 541.06 USD
77099-128 77099-130 77593-714
2-Fluoro-6-pyridone 97%
2-Fluoro-6-pyridone

Precaución: for Research Use Only

Formula: C₅H₄FNO
MW: 113.09 g/mol
MDL Number: MFCD03092938
CAS Number: 50543-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR