5-Bromo-2-iodoisopropylbenzene 97%

Supplier: Ambeed
Warning

A278089-1G A278089-250MG A278089-5G
77099-122EA 36 USD
77099-122 77099-124 77099-126
5-Bromo-2-iodoisopropylbenzene 97%
5-Bromo-2-iodoisopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrI
MDL Number: MFCD11845959
CAS Number: 1147014-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR