AGI-5198 ≥98%

Supplier: Ambeed
Warning

A276907-10MG A276907-1MG A276907-25MG A276907-2MG A276907-5MG
77099-100EA 14.86 USD
77099-100 77099-102 77099-104 77099-106 77099-108
AGI-5198 ≥98%
AGI-5198

Precaución: for Research Use Only

Formula: C₂₇H₃₁FN₄O₂
MDL Number: MFCD24848688
CAS Number: 1355326-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR