Methyl 3-(4-Iodophenyl)propanoate 97%

Supplier: Ambeed
Warning

A276800-1G A276800-250MG
77099-090EA 40 USD
77099-090 77099-092
Methyl 3-(4-Iodophenyl)propanoate 97%
Methyl 3-(4-Iodophenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁IO₂
MDL Number: MFCD18399500
CAS Number: 33994-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR