H-2-Abu-OtBu·HCl 98%

Supplier: Ambeed
Warning

Synonyms: H-Abu-OtBu.HCl, tert-Butyl (S)-2-aminobutyrate hydrochloride, H-Abu-OtBu·HCl

A276386-100G A276386-10G A276386-1G A276386-25G A276386-5G
77099-080EA 168 USD
77099-080 77099-082 77099-084 77099-086 77099-088
H-2-Abu-OtBu·HCl 98%
H-2-Abu-OtBu·HCl

Precaución: for Research Use Only

Formula: C₈H₁₈ClNO₂
MDL Number: MFCD00237697
CAS Number: 53956-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR