1-Phenylpyrrolidine 98%

Supplier: Ambeed
Warning

Synonyms: N-Phenylpyrrolidine

A275302-100G A275302-10G A275302-1G A275302-250MG A275302-25G A275302-5G
77099-068EA 455 USD
77099-068 77099-070 77099-072 77099-074 77099-076 77099-078
1-Phenylpyrrolidine 98%
1-Phenylpyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₃N
MDL Number: MFCD00015897
CAS Number: 4096-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR