(4-Ethoxy-2,5-difluorophenyl)boronic acid 95+%

Supplier: Ambeed
Warning

Synonyms: 2,5-Difluoro-4-ethoxyphenylboronic acid

A274571-1G A274571-250MG A274571-5G
77099-060EA 159.14 USD
77099-060 77099-062 77099-064
(4-Ethoxy-2,5-difluorophenyl)boronic acid 95+%
(4-Ethoxy-2,5-difluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BF₂O₃
MW: 201.96 g/mol
MDL Number: MFCD12026701
CAS Number: 1452575-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR