EPZ-015666 ≥98%

Supplier: Ambeed
Warning

A272770-10MG A272770-5MG
77099-040EA 27 USD
77099-040 77099-042
EPZ-015666 ≥98%
EPZ-015666

Precaución: for Research Use Only

Formula: C₂₀H₂₅N₅O₃
MDL Number: MFCD28411588
CAS Number: 1616391-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR