6-(Trifluoromethyl)-1H-indazol-5-amine 95%

Supplier: Ambeed
Warning

A272713-100MG A272713-250MG A272713-1G
77099-036EA 66.84 USD
77099-036 77099-038 77568-534
6-(Trifluoromethyl)-1H-indazol-5-amine 95%
6-(Trifluoromethyl)-1H-indazol-5-amine

Precaución: For research use only.

Formula: C₈H₆F₃N₃
MW: 201.14 g/mol
MDL Number: MFCD24642227
CAS Number: 1000373-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR