4-Ethoxy-2,3-difluoro-4'-(trans-4-propylcyclohexyl)biphenyl 97%

Supplier: Ambeed
Warning

Synonyms: 4-Ethoxy-2,3-difluoro-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl

A271860-1G A271860-25G A271860-5G
77099-010EA 7 USD
77099-010 77099-012 77099-014
4-Ethoxy-2,3-difluoro-4'-(trans-4-propylcyclohexyl)biphenyl 97%
4-Ethoxy-2,3-difluoro-4'-(trans-4-propylcyclohexyl)biphenyl

Precaución: for Research Use Only

Formula: C₂₃H₂₈F₂O
MW: 358.47 g/mol
MDL Number: MFCD18451991
CAS Number: 189750-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR