4-Bromo-2-chloroanisole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2-chloro-1-methoxybenzene

A271749-100G A271749-5G
77099-008EA 265.23 USD
77099-008 77571-678
4-Bromo-2-chloroanisole 97%
4-Bromo-2-chloroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrClO
MW: 221.4789 g/mol
MDL Number: MFCD00128076
CAS Number: 50638-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR