6-Chloroquinolin-2(1H)-one 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2-hydroxyquinoline

A271741-100G A271741-25G A271741-5G
77099-002EA 240 USD
77099-002 77099-004 77099-006
6-Chloroquinolin-2(1H)-one 98%
6-Chloroquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₆ClNO
MDL Number: MFCD02930633
CAS Number: 1810-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR