Di-tert-butyl 6-oxo-1,4-diazepane-1,4-dicarboxylate 95+%

Supplier: Ambeed
Warning

A271533-100MG A271533-1G A271533-250MG
77098-994EA 237.65 USD
77098-994 77098-996 77098-998
Di-tert-butyl 6-oxo-1,4-diazepane-1,4-dicarboxylate 95+%
Di-tert-butyl 6-oxo-1,4-diazepane-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₆N₂O₅
MW: 314.37 g/mol
MDL Number: MFCD21362344
CAS Number: 459417-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR