2-(Ethanesulfonyl)-5-nitropyridine 98%

Supplier: Ambeed
Warning

A270801-10G A270801-1G A270801-250MG A270801-5G
77098-986EA 200 USD
77098-986 77098-988 77098-990 77098-992
2-(Ethanesulfonyl)-5-nitropyridine 98%
2-(Ethanesulfonyl)-5-nitropyridine

Precaución: for Research Use Only

Formula: C₇H₈N₂O₄S
MDL Number: MFCD25121757
CAS Number: 767355-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR