2,2'-Dihydroxydiphenyl ether 98%

Supplier: Ambeed
Warning

Synonyms: Bis(2-hydroxyphenyl) Ether, 2,2'-Oxybisphenol, 2-Hydroxyphenyl Ether, 2,2'-Oxydiphenol

A267553-100MG A267553-250MG
77098-958EA 42.44 USD
77098-958 77098-960
2,2'-Dihydroxydiphenyl ether 98%
2,2'-Dihydroxydiphenyl ether

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₃
MW: 202.21 g/mol
MDL Number: MFCD00142736
CAS Number: 15764-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR