4-Fluorophenol 98%

Supplier: Ambeed
Warning

Synonyms: p-Fluorophenol, 1-Fluoro-4-hydroxybenzene

A267007-100G A267007-25G A267007-500G
77098-948EA 21.22 USD
77098-948 77098-950 77098-952
4-Fluorophenol 98%
4-Fluorophenol

Precaución: for Research Use Only

Formula: C₆H₅FO
MW: 112.1 g/mol
MDL Number: MFCD00002316
CAS Number: 371-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR