2-(Trimethylsilyl)ethyl-3-Nitro-1H-1,2,4-triazole-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 3-Nitro-1H-1,2,4-triazole-1-carboxylic Acid 2-(Trimethylsilyl)ethyl Ester, 3-Nitro-1-[2-(trimethylsilyl)ethoxycarbonyl]-1H-1,2,4-triazole, Teoc-NT, 2-(Trimethylsilyl)ethyl 3-Nitro-1H-1,2,4-triazole-1-carboxylate

A265949-1G A265949-5G A265949-250MG
77098-936EA 59.42 USD
77098-936 77098-938 77593-650
2-(Trimethylsilyl)ethyl-3-Nitro-1H-1,2,4-triazole-1-carboxylate 98%
2-(Trimethylsilyl)ethyl-3-Nitro-1H-1,2,4-triazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₄N₄O₄Si
MW: 258.31 g/mol
MDL Number: MFCD11042686
CAS Number: 1001067-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR