Z-D-Dab(Boc)-OH·DCHA 97%

Supplier: Ambeed
Warning

Synonyms: Z-D-Dab(Boc)-OH·DCHA, Dicyclohexylamine (R)-2-(((benzyloxy)carbonyl)amino)-4-((tert-butoxycarbonyl)amino)butanoate

A265246-1G A265246-250MG
77098-922EA 88.06 USD
77098-922 77098-924
Z-D-Dab(Boc)-OH·DCHA 97%
Z-D-Dab(Boc)-OH·DCHA

Precaución: for Research Use Only

Formula: C₂₉H₄₇N₃O₆
MW: 533.71 g/mol
MDL Number: MFCD01318750
CAS Number: 214852-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR