2,3-Difluoro-4-methoxyphenylacetonitrile 95%

Supplier: Ambeed
Warning

A265146-1G A265146-250MG A265146-5G
77098-916EA 39 USD
77098-916 77098-918 77098-920
2,3-Difluoro-4-methoxyphenylacetonitrile 95%
2,3-Difluoro-4-methoxyphenylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₇F₂NO
MDL Number: MFCD06660320
CAS Number: 886503-74-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR