2-Bromo-4-chloro-6-fluoroaniline 98%

Supplier: Ambeed
Warning

A264963-1G A264963-5G A264963-25G
77098-908EA 9 USD
77098-908 77098-910 77685-753
2-Bromo-4-chloro-6-fluoroaniline 98%
2-Bromo-4-chloro-6-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrClFN
MDL Number: MFCD00070746
CAS Number: 195191-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR