1-(tert-Butoxycarbonyl)-5-fluoropiperidine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A264786-100MG A264786-1G A264786-250MG
77098-890EA 126 USD
77098-890 77098-892 77098-894
1-(tert-Butoxycarbonyl)-5-fluoropiperidine-3-carboxylic acid 95%
1-(tert-Butoxycarbonyl)-5-fluoropiperidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₈FNO₄
MDL Number: MFCD17015060
CAS Number: 1241725-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR