5-Chloro-3-fluoropicolinaldehyde 98%

Supplier: Ambeed
Warning

A263827-250MG A263827-5G
77098-874EA 20 USD
77098-874 77098-876
5-Chloro-3-fluoropicolinaldehyde 98%
5-Chloro-3-fluoropicolinaldehyde

Precaución: for Research Use Only

Formula: C₆H₃ClFNO
MW: 159.54 g/mol
MDL Number: MFCD16606534
CAS Number: 214055-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR