3-Bromoimidazo[1,2-a]pyrimidine 98%

Supplier: Ambeed
Warning

A261216-1G A261216-250MG A261216-5G
77098-836EA 16 USD
77098-836 77098-838 77612-504
3-Bromoimidazo[1,2-a]pyrimidine 98%
3-Bromoimidazo[1,2-a]pyrimidine

Precaución: for Research Use Only

Formula: C6H4BrN3
MDL Number: MFCD06200962
CAS Number: 6840-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR