4-Bromo-3-fluorobenzenesulfonamide 98%

Supplier: Ambeed
Warning

A261205-1G A261205-25G A261205-5G
77098-830EA 11.67 USD
77098-830 77098-832 77098-834
4-Bromo-3-fluorobenzenesulfonamide 98%
4-Bromo-3-fluorobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₆H₅BrFNO₂S
MW: 254.08 g/mol
MDL Number: MFCD03094292
CAS Number: 263349-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR