5,5-Dimethyl-dihydro-furan-2-one 95%

Supplier: Ambeed
Warning

Synonyms: 5,5-Dimethyldihydrofuran-2(3H)-one

A261084-10G A261084-1G A261084-250MG A261084-5G
77098-822EA 759.61 USD
77098-822 77098-824 77098-826 77098-828
5,5-Dimethyl-dihydro-furan-2-one 95%
5,5-Dimethyl-dihydro-furan-2-one

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.142 g/mol
MDL Number: MFCD08061147
CAS Number: 3123-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR