1-(8-Bromoquinolin-4-yl)-N-methylmethanamine 95%

Supplier: Ambeed
Warning

A260178-100MG A260178-1G
77098-812EA 91.24 USD
77098-812 77098-814
1-(8-Bromoquinolin-4-yl)-N-methylmethanamine 95%
1-(8-Bromoquinolin-4-yl)-N-methylmethanamine

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrN₂
MDL Number: MFCD12024576
CAS Number: 1190322-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR