3-Bromo-2-(trifluoromethyl)aniline 95%
Supplier: Ambeed
|
Warning
|
A259990-100MG
A259990-1G
A259990-250MG
A259990-5G
77098-800EA
73
USD
77098-800
77098-802
77098-804
77098-806
3-Bromo-2-(trifluoromethyl)aniline 95%
3-Bromo-2-(trifluoromethyl)aniline
Precaución: for Research Use Only
Formula:
C₇H₅BrF₃N MW: 240.021 g/mol |
MDL Number:
MFCD13194482 CAS Number: 244246-71-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...