2,4-Dimethyl-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A259259-1G A259259-250MG
77098-790EA 559.1 USD
77098-790 77098-792
2,4-Dimethyl-1H-pyrrolo[2,3-b]pyridine 95%
2,4-Dimethyl-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₉H₁₀N₂
MW: 146.18 g/mol
MDL Number: MFCD03425552
CAS Number: 1190313-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR