4-Bromo-N-ethyl-3-methylbenzenesulfonamide 95+%

Supplier: Ambeed
Warning

A259078-1G A259078-250MG A259078-5G
77098-784EA 125.19 USD
77098-784 77098-786 77098-788
4-Bromo-N-ethyl-3-methylbenzenesulfonamide 95+%
4-Bromo-N-ethyl-3-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₉H₁₂BrNO₂S
MW: 278.16 g/mol
MDL Number: MFCD11618328
CAS Number: 1094563-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR