1-Isobutyl-2-nitrobenzene 95+%

Supplier: Ambeed
Warning

A257443-1G A257443-250MG A257443-5G
77098-756EA 46 USD
77098-756 77098-758 77901-278
1-Isobutyl-2-nitrobenzene 95+%
1-Isobutyl-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00130031
CAS Number: 19370-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR