2-Fluoro-5-(trifluoromethyl)benzene sulfonamide 95%

Supplier: Ambeed
Warning

A255357-100MG A255357-1G A255357-250MG A255357-5G
77098-728EA 31.83 USD
77098-728 77098-730 77098-732 77098-734
2-Fluoro-5-(trifluoromethyl)benzene sulfonamide 95%
2-Fluoro-5-(trifluoromethyl)benzene sulfonamide

Precaución: for Research Use Only

Formula: C₇H₅F₄NO₂S
MW: 243.18 g/mol
MDL Number: MFCD12026239
CAS Number: 217454-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR