N(α)-Boc-D-histidine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-D-His-OH
A252722-10G
A252722-250MG
A252722-1G
A252722-5G
77098-716EA
84
USD
77098-716
77098-718
77685-718
77685-719
N(α)-Boc-D-histidine 98%
N(α)-Boc-D-histidine
Precaución: for Research Use Only
Formula:
C₁₁H₁₇N₃O₄ MW: 255.3 g/mol |
MDL Number:
MFCD00037851 CAS Number: 50654-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...