1-Methyl-2-(methylsulfonyl)-5-nitro-1H-imidazole 96%

Supplier: Ambeed
Warning

A252418-100MG A252418-250MG
77098-712EA 18.04 USD
77098-712 77098-714
1-Methyl-2-(methylsulfonyl)-5-nitro-1H-imidazole 96%
1-Methyl-2-(methylsulfonyl)-5-nitro-1H-imidazole

Precaución: for Research Use Only

Formula: C₅H₇N₃O₄S
MW: 205.19 g/mol
MDL Number: MFCD26407320
CAS Number: 1615-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR