6-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid 98%
Supplier: Ambeed
|
Warning
|
A252397-100G
A252397-10G
A252397-1G
A252397-250MG
A252397-25G
77098-702EA
2517
USD
77098-702
77098-704
77098-706
77098-708
77098-710
6-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid 98%
6-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only
Formula:
C₈H₃BrF₄O₂ MW: 287.006 g/mol |
MDL Number:
MFCD21365092 CAS Number: 1026962-68-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...