4,4,5,5-Tetramethyl-2-(2-methyl-4-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Methyl-4-(trifluoromethyl)phenylboronic acid pinacol ester
A252157-1G
A252157-250MG
A252157-5G
77098-696EA
68.96
USD
77098-696
77098-698
77098-700
4,4,5,5-Tetramethyl-2-(2-methyl-4-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane 98%
4,4,5,5-Tetramethyl-2-(2-methyl-4-(trifluoromethyl)phenyl)-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₄H₁₈BF₃O₂ MW: 286.1 g/mol |
MDL Number:
MFCD18729901 CAS Number: 1689509-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...