2-Methyl-5-(trifluoromethoxy)aniline 97%

Supplier: Ambeed
Warning

A251284-5G A251284-1G A251284-250MG
77098-688EA 204 USD
77098-688 77900-769 77901-251
2-Methyl-5-(trifluoromethoxy)aniline 97%
2-Methyl-5-(trifluoromethoxy)aniline

Precaución: for Research Use Only

Formula: C₈H₈F₃NO
MDL Number: MFCD07777196
CAS Number: 933674-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR