2-Chloro-5-methoxy-4-nitrophenol 95%

Supplier: Ambeed
Danger

A250450-100MG A250450-1G
77098-674EA 51.99 USD
77098-674 77098-676
2-Chloro-5-methoxy-4-nitrophenol 95%
2-Chloro-5-methoxy-4-nitrophenol

Precaución: for Research Use Only

Formula: C₇H₆ClNO₄
MW: 203.57 g/mol
MDL Number: MFCD18431954
CAS Number: 100959-51-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR