Moxifloxacin hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A250011-1G
A250011-25G
A250011-50MG
A250011-5G
A250011-10G
A250011-1MG
A250011-250MG
A250011-100G
77098-660EA
16
USD
77098-660
77098-662
77098-664
77568-430
77612-412
77685-707
77685-708
77901-249
Moxifloxacin hydrochloride 98%
Moxifloxacin hydrochloride
Precaución: For research use only.
Formula:
C₂₁H₂₅ClFN₃O₄ |
MDL Number:
MFCD00949117 CAS Number: 186826-86-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...