2-(Ethylsulfonyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%

Supplier: Ambeed
Warning

A249208-1G A249208-250MG A249208-5G
77098-644EA 160.2 USD
77098-644 77098-646 77098-648
2-(Ethylsulfonyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98%
2-(Ethylsulfonyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₄S
MW: 297.18 g/mol
MDL Number: MFCD19684130
CAS Number: 1449010-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR