Boc-β-Ala-OSu 95%

Supplier: Ambeed
Warning

A249016-1G A249016-25G
77098-642EA 10.61 USD
77098-642 77612-402
Boc-β-Ala-OSu 95%
Boc-Beta-Ala-OSu

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₆
MW: 286.284 g/mol
MDL Number: MFCD00038647
CAS Number: 32703-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR