(S)-1-(Naphthalen-1-yl)ethanol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-1-(1-Hydroxyethyl)naphthalene, (S)-(-)-alpha-Methyl-1-naphthalenemethanol, (S)-(-)-1-(1-Naphthyl)ethanol

A248873-10G A248873-1G A248873-250MG A248873-25G A248873-5G
77098-620EA 176.11 USD
77098-620 77098-622 77098-624 77098-626 77098-628
(S)-1-(Naphthalen-1-yl)ethanol 97%
(S)-1-(Naphthalen-1-yl)ethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₂O
MW: 172.23 g/mol
MDL Number: MFCD00077831
CAS Number: 15914-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR